siRNA Details
| Gene Symbol: HDAC11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136041.2 | NP_001129513.1 | 9 | 796 | ||
| NM_024827.3 | NP_079103.2 | 9 | 897 | |||
| XM_005265491.1 | XP_005265548.1 | 9 | 862 | |||
| XM_005265492.1 | XP_005265549.1 | 6 | 704 | |||
| XM_006713338.1 | XP_006713401.1 | 6 | 660 | |||
| XM_011534131.1 | XP_011532433.1 | 9 | 985 | |||
| XM_011534132.2 | XP_011532434.2 | 6 | 550 | |||
| XM_011534135.1 | XP_011532437.1 | 8 | 704 | |||
| XM_017007234.1 | XP_016862723.1 | 6 | 748 | |||
| GenBank mRNA | AK025426.1 | - | 9 | 788 | ||
| AK025890.1 | - | 9 | 788 | |||
| AK293223.1 | - | 9 | 729 | |||
| AK299021.1 | - | 5 | 468 | |||
| AL137362.1 | - | 6 | 431 | |||
| AL834223.1 | - | 8 | 1112 | |||
| BC009676.1 | - | 8 | 773 | 3906049 | ||
| CR457341.1 | - | 5 | 398 | |||

HDAC11
Genomic Map