siRNA Details
Gene Symbol: GGNBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024835.4 | NP_079111.1 | 14 | 2321 | ||
XM_005257689.2 | XP_005257746.1 | 14 | 2303 | |||
XM_011525265.2 | XP_011523567.1 | 13 | 2066 | |||
XM_011525266.1 | XP_011523568.1 | 13 | 2189 | |||
XM_017025107.1 | XP_016880596.1 | 13 | 2034 | |||
GenBank mRNA | AF126964.1 | - | 13 | 2052 | ||
AJ404670.2 | - | 6 | 878 | |||
AK026214.1 | - | 6 | 955 | |||
AK290687.1 | - | 13 | 2077 | |||
AK301614.1 | - | 11 | 1609 | |||
BC137491.1 | - | 14 | 2187 | 9021491 |
