siRNA Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001031721.3 | NP_001026891.2 | 6 | 1955 | ||
| NM_024840.3 | NP_079116.2 | 6 | 1847 | |||
| XM_005259269.3 | XP_005259326.1 | 5 | 2061 | |||
| XM_011527333.2 | XP_011525635.1 | 6 | 2171 | |||
| XM_017027315.1 | XP_016882804.1 | 5 | 2019 | |||
| GenBank mRNA | AK023652.1 | - | 1 | 652 | ||
| AK027565.1 | - | 6 | 1847 | |||
| BC057776.1 | - | 6 | 1914 | 30338140 | ||
| CA427456.1 | - | 1 | 397 | |||

ZNF613
Genomic Map