siRNA Details
| Gene Symbol: PYROXD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024854.3 | NP_079130.2 | 12 | 1499 | ||
| XM_017019976.1 | XP_016875465.1 | 6 | 813 | |||
| GenBank mRNA | AK025681.1 | - | 12 | 1402 | ||
| AK125461.1 | - | 12 | 1912 | |||
| AK307710.1 | - | 11 | 1324 | |||
| AL832441.1 | - | 12 | 1426 | |||
| BC021662.1 | - | 12 | 1400 | 3826938 | ||

PYROXD1
Genomic Map