siRNA Details
| Gene Symbol: ATAD5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024857.4 | NP_079133.3 | 9 | 3316 | ||
| XM_011525269.2 | XP_011523571.1 | 9 | 3337 | |||
| XM_011525270.2 | XP_011523572.1 | 9 | 3337 | |||
| XM_011525273.2 | XP_011523575.1 | 9 | 4354 | |||
| XM_011525274.2 | XP_011523576.1 | 4 | 565 | |||
| XM_017025111.1 | XP_016880600.1 | 9 | 3312 | |||
| GenBank mRNA | AJ314648.2 | - | 9 | 3284 | ||
| AK022797.1 | - | 8 | 1811 | |||
| AL832103.1 | - | 9 | 3317 | |||
| AY557611.1 | - | 9 | 3284 | |||

ATAD5
Genomic Map