siRNA Details
| Gene Symbol: C2orf54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085437.2 | NP_001078906.2 | 5 | 1452 | ||
| NM_001282921.1 | NP_001269850.1 | 5 | 945 | |||
| NM_024861.3 | NP_079137.3 | 5 | 888 | |||
| XM_011511877.1 | XP_011510179.1 | 6 | 1444 | |||
| GenBank mRNA | AK026324.1 | - | 5 | 885 | ||
| AK056601.1 | - | 5 | 1452 | |||
| AK096434.1 | - | 5 | 945 | |||
| BC125154.1 | - | 5 | 876 | 40120634 | ||
| BC125155.1 | - | 5 | 876 | 40120635 | ||

C2orf54
Genomic Map