siRNA Details
| Gene Symbol: SYNPO2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001114133.2 | NP_001107605.1 | 4 | 2500 | ||
| NM_024875.4 | NP_079151.2 | 2 | 1677 | |||
| XM_005270158.2 | XP_005270215.1 | 2 | 1999 | |||
| XM_005270159.3 | XP_005270216.1 | 2 | 1633 | |||
| GenBank mRNA | AB188489.1 | - | 4 | 2469 | ||
| AK022983.1 | - | 2 | 1667 | |||
| AK295873.1 | - | 4 | 2389 | |||
| BC142635.1 | - | 2 | 1659 | 40147769 | ||
| BX502518.1 | - | 1 | 327 | |||

SYNPO2L
Genomic Map