siRNA Details
| Gene Symbol: GRHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024915.3 | NP_079191.2 | 5 | 1041 | ||
| XM_011517305.2 | XP_011515607.1 | 5 | 1072 | |||
| XM_011517306.2 | XP_011515608.1 | 5 | 809 | |||
| XM_011517307.2 | XP_011515609.1 | 5 | 1039 | |||
| GenBank mRNA | AK023844.1 | - | 5 | 723 | ||
| AK292853.1 | - | 5 | 723 | |||
| AK296817.1 | - | 5 | 1040 | |||
| BC069618.1 | - | 5 | 723 | 7262389 | ||
| BC069633.1 | - | 5 | 783 | 7262401 | ||
| BC069638.1 | - | 5 | 723 | 7262413 | ||
| BC129822.1 | - | 5 | 723 | 40113571 | ||
| BC129823.1 | - | 5 | 723 | 40113576 | ||

GRHL2
Genomic Map