siRNA Details
| Gene Symbol: CES3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001185176.1 | NP_001172105.1 | 6 | 927 | ||
| NM_001185177.1 | NP_001172106.1 | 13 | 1634 | |||
| NM_024922.5 | NP_079198.2 | 13 | 1643 | |||
| XM_011522975.2 | XP_011521277.1 | 13 | 1715 | |||
| GenBank mRNA | AK000105.1 | - | 13 | 1582 | ||
| AK025158.1 | - | 3 | 225 | |||
| AK025389.1 | - | 6 | 926 | |||
| AK097538.1 | - | 12 | 2085 | |||
| AY358609.1 | - | 13 | 1569 | |||
| BC053670.1 | - | 13 | 1618 | 6145696 | ||

CES3
Genomic Map