siRNA Details
| Gene Symbol: ANKRD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115116.1 | NP_001108588.1 | 5 | 1090 | ||
| NM_024933.3 | NP_079209.3 | 5 | 1090 | |||
| XM_005264569.4 | XP_005264626.1 | 5 | 2250 | |||
| XM_011533114.2 | XP_011531416.1 | 4 | 761 | |||
| GenBank mRNA | AK093479.1 | - | 5 | 926 | ||
| BC035234.1 | - | 5 | 1090 | 5166484 | ||

ANKRD53
Genomic Map