siRNA Details
| Gene Symbol: ANKRD53 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001115116.1 | NP_001108588.1 | 3 | 682 | ||
| NM_024933.3 | NP_079209.3 | 3 | 682 | |||
| XM_005264569.4 | XP_005264626.1 | 3 | 1842 | |||
| XM_011533114.2 | XP_011531416.1 | 3 | 518 | |||
| GenBank mRNA | AK022118.1 | - | 1 | 1093 | ||
| AK093479.1 | - | 3 | 518 | |||
| BC035234.1 | - | 3 | 682 | 5166484 | ||
| DB036445.1 | - | 2 | 280 | |||

ANKRD53
Genomic Map