siRNA Details
| Gene Symbol: RMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024945.2 | NP_079221.2 | 3 | 943 | ||
| XM_005252211.2 | XP_005252268.1 | 3 | 812 | |||
| XM_005252213.2 | XP_005252270.1 | 4 | 888 | |||
| XM_011519034.2 | XP_011517336.1 | 4 | 1001 | |||
| XM_017015140.1 | XP_016870629.1 | 3 | 753 | |||
| GenBank mRNA | AK022950.1 | - | 3 | 867 | ||
| BC020606.1 | - | 3 | 765 | |||
| BC032494.1 | - | 4 | 879 | |||
| BC039999.2 | - | 3 | 770 | 5554889 | ||
| BC053549.1 | - | 3 | 703 | |||
| BC064937.1 | - | 2 | 683 | 6158300 | ||

RMI1
Genomic Map