siRNA Details
| Gene Symbol: RMI1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024945.2 | NP_079221.2 | 3 | 1055 | ||
| XM_005252211.2 | XP_005252268.1 | 3 | 924 | |||
| XM_005252213.2 | XP_005252270.1 | 4 | 1000 | |||
| XM_011519034.2 | XP_011517336.1 | 4 | 1113 | |||
| XM_017015140.1 | XP_016870629.1 | 3 | 865 | |||
| GenBank mRNA | AK022950.1 | - | 3 | 979 | ||
| BC020606.1 | - | 3 | 877 | |||
| BC032494.1 | - | 4 | 991 | |||
| BC039999.2 | - | 3 | 882 | 5554889 | ||
| BC053549.1 | - | 3 | 815 | |||
| BC064937.1 | - | 2 | 795 | 6158300 | ||

RMI1
Genomic Map