siRNA Details
| Gene Symbol: FBXL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001321213.1 | NP_001308142.1 | 3 | 1051 | ||
| NM_024963.5 | NP_079239.3 | 3 | 1051 | |||
| XM_011515531.2 | XP_011513833.1 | 3 | 1066 | |||
| XM_011515532.2 | XP_011513834.1 | 3 | 1066 | |||
| GenBank mRNA | AK057042.1 | - | 3 | 1005 | ||
| AK095394.1 | - | 1 | 578 | |||
| AK294949.1 | - | 3 | 1045 | |||
| BC006426.1 | - | 3 | 1021 | 4026501 | ||
| BC024256.1 | - | 3 | 1021 | 3954975 | ||

FBXL18
Genomic Map