siRNA Details
| Gene Symbol: ARL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025047.2 | NP_079323.1 | 1 | 255 | ||
| GenBank mRNA | AK026248.1 | - | 1 | 256 | ||
| BC034354.1 | - | 1 | 238 | 4747382 | ||
| BM760777.1 | - | 1 | 256 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ARL14| Gene Symbol: ARL14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025047.2 | NP_079323.1 | 1 | 255 | ||
| GenBank mRNA | AK026248.1 | - | 1 | 256 | ||
| BC034354.1 | - | 1 | 238 | 4747382 | ||
| BM760777.1 | - | 1 | 256 | |||