siRNA Details
| Gene Symbol: CCDC170 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025059.3 | NP_079335.2 | 3 | 294 | ||
| XM_011536147.2 | XP_011534449.1 | 3 | 332 | |||
| XM_011536148.2 | XP_011534450.1 | 3 | 335 | |||
| GenBank mRNA | AK026958.1 | - | 3 | 207 | ||
| AK225330.1 | - | 3 | 208 | |||
| BC060803.1 | - | 3 | 243 | 5298914 | ||
| BI562667.1 | - | 3 | 248 | |||
| DB232703.1 | - | 3 | 294 | |||

CCDC170
Genomic Map