siRNA Details
| Gene Symbol: LRRC8E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001268284.2 | NP_001255213.1 | 4 | 676 | ||
| NM_001268285.2 | NP_001255214.1 | 2 | 390 | |||
| NM_025061.5 | NP_079337.2 | 3 | 533 | |||
| XM_011528319.2 | XP_011526621.1 | 3 | 1261 | |||
| GenBank mRNA | AK091134.1 | - | 3 | 533 | ||
| BC070089.1 | - | 3 | 533 | 30346745 | ||
| BX538180.1 | - | 3 | 523 | |||
| CD109368.1 | - | 3 | 560 | |||
| CF593782.1 | - | 4 | 659 | |||
| DA447942.1 | - | 2 | 355 | |||

LRRC8E
Genomic Map