siRNA Details
| Gene Symbol: EFHC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025184.3 | NP_079460.2 | 5, 6 | 936 | ||
| XM_006724562.2 | XP_006724625.1 | 4, 5 | 649 | |||
| GenBank mRNA | AK026496.1 | - | 5, 6 | 789 | ||
| AK291187.1 | - | 5, 6 | 1077 | |||
| AK292967.1 | - | 5, 6 | 920 | |||
| AK297714.1 | - | 5, 6 | 659 | |||
| BC031039.1 | - | 5, 6 | 931 | 5272398 | ||
| CR749363.1 | - | 1, 2 | 205 | |||

EFHC2
Genomic Map