siRNA Details
| Gene Symbol: WDCP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142319.1 | NP_001135791.1 | 2 | 232 | ||
| NM_025203.2 | NP_079479.1 | 2 | 232 | |||
| XM_017005029.1 | XP_016860518.1 | 2 | 227 | |||
| GenBank mRNA | AK025598.1 | - | 2 | 159 | ||
| BC035698.2 | - | 2 | 167 | 5547301 | ||
| DB292134.1 | - | 2 | 234 | |||

WDCP
Genomic Map