siRNA Details
| Gene Symbol: SPHKAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142644.1 | NP_001136116.1 | 7 | 4167 | ||
| NM_030623.3 | NP_085126.2 | 7 | 4167 | |||
| XM_005246870.4 | XP_005246927.1 | 8 | 4639 | |||
| XM_006712777.3 | XP_006712840.1 | 8 | 4600 | |||
| XM_006712778.3 | XP_006712841.1 | 8 | 4599 | |||
| XM_011511925.2 | XP_011510227.1 | 9 | 4718 | |||
| GenBank mRNA | AK090876.1 | - | 1 | 1242 | ||
| AK095042.1 | - | 1 | 329 | |||
| AK127071.1 | - | 7 | 4217 | |||
| BC104954.1 | - | 7 | 4167 | 8143957 | ||
| BC104956.1 | - | 7 | 4167 | 8143959 | ||
| BC143408.1 | - | 7 | 4167 | |||
| CR749297.1 | - | 1 | 3273 | |||
| CR749494.1 | - | 7 | 4149 | |||

SPHKAP
Genomic Map