siRNA Details
| Gene Symbol: ESX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153448.3 | NP_703149.1 | 1 | 138 | ||
| GenBank mRNA | AY114148.1 | - | 1 | 112 | ||
| BC042633.1 | - | 1 | 99 | 5170565 | ||
| BC053599.1 | - | 1 | 127 | 5742902 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ESX1| Gene Symbol: ESX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_153448.3 | NP_703149.1 | 1 | 138 | ||
| GenBank mRNA | AY114148.1 | - | 1 | 112 | ||
| BC042633.1 | - | 1 | 99 | 5170565 | ||
| BC053599.1 | - | 1 | 127 | 5742902 | ||