siRNA Details
| Gene Symbol: ACAD10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136538.1 | NP_001130010.1 | 21 | 3268 | ||
| NM_025247.5 | NP_079523.3 | 20 | 3175 | |||
| GenBank mRNA | AK097425.1 | - | 13 | 1901 | ||
| AL832043.1 | - | 21 | 3246 | |||
| AY323912.1 | - | 20 | 3131 | |||
| BC003698.1 | - | 16 | 2332 | |||
| BC126358.1 | - | 20 | 3089 | 8992074 | ||

ACAD10
Genomic Map