siRNA Details
| Gene Symbol: KIAA1683 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001145304.1 | NP_001138776.1 | 3 | 1630 | ||
| NM_001145305.1 | NP_001138777.1 | 3 | 1669 | |||
| NM_025249.3 | NP_079525.1 | 3 | 1630 | |||
| XM_005260084.1 | XP_005260141.1 | 3 | 1555 | |||
| XM_011528324.1 | XP_011526626.1 | 2 | 1576 | |||
| GenBank mRNA | AK302431.1 | - | 3 | 1582 | ||
| AL136867.1 | - | 3 | 1547 | |||
| BC112932.1 | - | 3 | 1556 | 5744104 | ||
| BC127099.1 | - | 2 | 1496 | |||

KIAA1683
Genomic Map