siRNA Details
| Gene Symbol: PRR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077497.2 | NP_001070965.1 | 2 | 754 | ||
| NM_025263.3 | NP_079539.2 | 3 | 817 | |||
| GenBank mRNA | AK074531.1 | - | 2 | 183 | ||
| AK131382.1 | - | 1 | 2145 | |||
| AK298621.1 | - | 3 | 541 | |||
| AK316586.1 | - | 3 | 546 | |||
| BC126455.1 | - | 3 | 397 | 8992171 | ||
| CN258740.1 | - | 3 | 210 | |||
| DR762096.1 | - | 2 | 144 | |||
| U63336.1 | - | 3 | 640 | |||

PRR3
Genomic Map