siRNA Details
| Gene Symbol: PTGES3L-AARSD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136042.2 | NP_001129514.2 | 3 | 619 | ||
| NM_025267.3 | NP_079543.1 | 3 | 235 | |||
| GenBank mRNA | AF039235.1 | - | 3 | 324 | ||
| AK094084.1 | - | 3 | 173 | |||
| AK295447.1 | - | 3 | 619 | |||
| BC004172.2 | - | 3 | 235 | 2823004 | ||
| BC019324.1 | - | 3 | 235 | 2823004 | ||
| Gene Symbol: PTGES3L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001142653.1 | NP_001136125.1 | 3 | 619 | ||
| NM_001142654.1 | NP_001136126.1 | 2 | 505 | |||
| NM_001261430.1 | NP_001248359.1 | 3 | 619 | |||
| GenBank mRNA | AK056965.1 | - | 2 | 61 | ||
| AK093402.1 | - | 1 | 929 | |||
| DA900052.1 | - | 3 | 172 | |||

PTGES3L-AARSD1
Genomic Map