siRNA Details
| Gene Symbol: THAP7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008695.1 | NP_001008695.1 | 3 | 219 | ||
| NM_030573.2 | NP_085050.2 | 2 | 393 | |||
| GenBank mRNA | AK024609.1 | - | 1 | 489 | ||
| AK025906.1 | - | 1 | 257 | |||
| AK315457.1 | - | 2 | 378 | |||
| BC004346.1 | - | 2 | 364 | 3633743 | ||
| BE264280.1 | - | 2 | 292 | |||
| BI223500.1 | - | 2 | 336 | |||
| BM790105.1 | - | 3 | 219 | |||
| CR456343.1 | - | 2 | 211 | |||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_026909, AK154641, BC018221, AK007329.

THAP7
Genomic Map