siRNA Details
| Gene Symbol: TMEM177 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001105198.1 | NP_001098668.1 | 2 | 693 | ||
| NM_001105199.1 | NP_001098669.1 | 2 | 301 | |||
| NM_030577.2 | NP_085054.2 | 2 | 352 | |||
| GenBank mRNA | AK096890.1 | - | 2 | 298 | ||
| AK130088.1 | - | 2 | 301 | |||
| AK299078.1 | - | 2 | 298 | |||
| AK308503.1 | - | 2 | 316 | |||
| BC004404.1 | - | 2 | 340 | 3637432 | ||
| BG280893.1 | - | 2 | 295 | |||
| BG776153.1 | - | 2 | 323 | |||

TMEM177
Genomic Map