siRNA Details
| Gene Symbol: CMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030629.2 | NP_085132.1 | 14 | 1418 | ||
| NM_198390.2 | NP_938204.2 | 14 | 1644 | |||
| XM_005256179.4 | XP_005256236.4 | 14 | 2599 | |||
| XM_005256181.2 | XP_005256238.1 | 13 | 1375 | |||
| XM_005256182.1 | XP_005256239.1 | 12 | 1132 | |||
| XM_011523352.1 | XP_011521654.1 | 14 | 1604 | |||
| XM_011523353.1 | XP_011521655.1 | 12 | 1512 | |||
| XM_017023733.1 | XP_016879222.1 | 12 | 5989 | |||
| GenBank mRNA | AB051481.1 | - | 14 | 1525 | ||
| AK096598.1 | - | 14 | 1605 | |||
| AK304418.1 | - | 13 | 1380 | |||
| AY172689.1 | - | 14 | 1418 | |||
| BC038113.1 | - | 12 | 1341 | 4818904 | ||

CMIP
Genomic Map