siRNA Details
| Gene Symbol: CMIP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030629.2 | NP_085132.1 | 18, 19 | 1926 | ||
| NM_198390.2 | NP_938204.2 | 18, 19 | 2152 | |||
| XM_005256179.4 | XP_005256236.4 | 18, 19 | 3107 | |||
| XM_005256181.2 | XP_005256238.1 | 17, 18 | 1883 | |||
| XM_005256182.1 | XP_005256239.1 | 16, 17 | 1640 | |||
| XM_011523352.1 | XP_011521654.1 | 17, 18 | 2064 | |||
| XM_011523353.1 | XP_011521655.1 | 16, 17 | 2020 | |||
| XM_017023733.1 | XP_016879222.1 | 16, 17 | 6497 | |||
| GenBank mRNA | AB051481.1 | - | 18, 19 | 2033 | ||
| AK096598.1 | - | 18, 19 | 2113 | |||
| AK304418.1 | - | 17, 18 | 1888 | |||
| AY172689.1 | - | 18, 19 | 1926 | |||
| BC038113.1 | - | 16, 17 | 1849 | 4818904 | ||

CMIP
Genomic Map