siRNA Details
| Gene Symbol: ZNF436 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001077195.1 | NP_001070663.1 | 4 | 961 | ||
| NM_030634.2 | NP_085137.1 | 3 | 783 | |||
| XM_011542201.1 | XP_011540503.1 | 3 | 865 | |||
| GenBank mRNA | AB051497.1 | - | 4 | 539 | ||
| AK092364.1 | - | 3 | 735 | |||
| AK308173.1 | - | 3 | 480 | |||
| AL834485.1 | - | 3 | 783 | |||
| BC056400.1 | - | 3 | 694 | 5244536 | ||

ZNF436
Genomic Map