siRNA Details
| Gene Symbol: LNPK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001305008.1 | NP_001291937.1 | 8 | 725 | ||
| NM_001305009.1 | NP_001291938.1 | 8 | 720 | |||
| NM_001305010.1 | NP_001291939.1 | 7 | 533 | |||
| NM_001305011.1 | NP_001291940.1 | 6 | 490 | |||
| NM_030650.2 | NP_085153.1 | 8 | 720 | |||
| NR_130941.1 | 9 | 1009 | ||||
| XM_005246878.3 | XP_005246935.1 | 8 | 656 | |||
| XM_005246879.4 | XP_005246936.1 | 8 | 667 | |||
| XM_005246880.3 | XP_005246937.1 | 4 | 357 | |||
| XM_006712783.2 | XP_006712846.1 | 8 | 862 | |||
| XM_011511945.2 | XP_011510247.1 | 9 | 1071 | |||
| XM_017005055.1 | XP_016860544.1 | 4 | 265 | |||
| GenBank mRNA | AB051502.1 | - | 8 | 626 | ||
| AK056532.1 | - | 8 | 643 | |||
| AK302833.1 | - | 7 | 467 | |||
| AL832947.1 | - | 5 | 383 | |||
| BC105132.1 | - | 8 | 650 | 8144135 | ||
| BC105134.1 | - | 8 | 650 | 8144137 | ||
| BC110329.1 | - | 6 | 419 | 5732809 | ||
| BC143681.1 | - | 8 | 650 | 9052195 | ||
| BC143683.1 | - | 7 | 463 | |||
| CR936742.1 | - | 8 | 669 | |||

LNPK
Genomic Map