siRNA Details
| Gene Symbol: SLC25A32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030780.4 | NP_110407.2 | 3 | 771 | ||
| NR_102338.1 | 4 | 880 | ||||
| GenBank mRNA | AF283645.1 | - | 3 | 501 | ||
| AK027531.1 | - | 3 | 540 | |||
| AK027787.1 | - | 3 | 505 | |||
| AL046268.1 | - | 3 | 341 | |||
| BC021893.1 | - | 3 | 479 | 4524907 | ||

SLC25A32
Genomic Map