siRNA Details
| Gene Symbol: OR51G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005238.1 | NP_001005238.1 | 1 | 769 | ||
| GenBank mRNA | BC136830.1 | - | 1 | 798 | 9020821 | |
| BC136831.1 | - | 1 | 798 | 9020822 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

OR51G2| Gene Symbol: OR51G2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001005238.1 | NP_001005238.1 | 1 | 769 | ||
| GenBank mRNA | BC136830.1 | - | 1 | 798 | 9020821 | |
| BC136831.1 | - | 1 | 798 | 9020822 | ||