siRNA Details
| Gene Symbol: OR51E2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030774.3 | NP_110401.1 | 2 | 271 | ||
| GenBank mRNA | AF369708.1 | - | 2 | 177 | ||
| AK314051.1 | - | 2 | 271 | |||
| AY033942.1 | - | 2 | 268 | |||
| BC020768.1 | - | 3 | 144 | 4249775 | ||

OR51E2
Genomic Map