siRNA Details
| Gene Symbol: SGPP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030791.3 | NP_110418.1 | 3 | 1178 | ||
| XM_017021678.1 | XP_016877167.1 | 4 | 1242 | |||
| GenBank mRNA | AF349315.1 | - | 3 | 1100 | ||
| AJ293294.1 | - | 3 | 1059 | |||
| AK314188.1 | - | 3 | 1124 | |||
| BC063839.1 | - | 3 | 1121 | 6184825 | ||
| BC070060.1 | - | 3 | 1138 | 30347615 | ||

SGPP1
Genomic Map