siRNA Details
| Gene Symbol: C1orf21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030806.3 | NP_110433.1 | 5 | 759 | ||
| GenBank mRNA | AF312864.1 | - | 5 | 681 | ||
| AK056487.1 | - | 5 | 495 | |||
| AK312062.1 | - | 4 | 396 | |||
| AY258286.1 | - | 5 | 672 | |||
| BC028567.2 | - | 5 | 717 | 3635521 | ||
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_197990, AK148738, BC034723, AK006337, BC055912, and others.

C1orf21
Genomic Map