siRNA Details
| Gene Symbol: TRIM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030912.2 | NP_112174.2 | 1 | 675 | ||
| GenBank mRNA | AF220034.1 | - | 1 | 552 | ||
| AF281046.1 | - | 1 | 561 | |||
| AL360160.1 | - | 1 | 561 | |||
| BC021925.1 | - | 1 | 674 | 4857393 | ||
| BG746631.1 | - | 1 | 675 | |||

TRIM8
Genomic Map