siRNA Details
| Gene Symbol: SHCBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030933.2 | NP_112195.2 | 7 | 1531 | ||
| GenBank mRNA | AF288397.1 | - | 8 | 1189 | ||
| AF288398.1 | - | 7 | 1530 | |||
| BC026084.1 | - | 6 | 1711 | |||
| BC050305.1 | - | 7 | 1483 | 4837018 | ||
| BC132764.1 | - | 7 | 1507 | 40146786 | ||
| BC144326.1 | - | 7 | 1503 | |||

SHCBP1L
Genomic Map