siRNA Details
| Gene Symbol: SHCBP1L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030933.2 | NP_112195.2 | 10 | 2232 | ||
| GenBank mRNA | AA448460.1 | - | 1 | 148 | ||
| AF288397.1 | - | 11 | 1890 | |||
| AF288398.1 | - | 10 | 2231 | |||
| BC026084.1 | - | 9 | 2412 | |||
| BC050305.1 | - | 10 | 2184 | 4837018 | ||
| BC132764.1 | - | 10 | 2208 | 40146786 | ||
| BC144326.1 | - | 10 | 2204 | |||

SHCBP1L
Genomic Map