siRNA Details
Gene Symbol: NUAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030952.2 | NP_112214.2 | 5 | 760 | ||
GenBank mRNA | AK074830.1 | - | 5 | 714 | ||
AK303577.1 | - | 5 | 760 | |||
AL136891.1 | - | 5 | 724 | |||
BC017306.2 | - | 5 | 696 | 5088037 | ||
BX537564.1 | - | 1 | 2256 | |||
CR749209.1 | - | 1 | 4189 |
