siRNA Details
Gene Symbol: NUAK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030952.2 | NP_112214.2 | 6 | 928 | ||
XM_005245515.4 | XP_005245572.1 | 2 | 185 | |||
GenBank mRNA | AK074830.1 | - | 6 | 882 | ||
AK303577.1 | - | 6 | 928 | |||
AL136891.1 | - | 6 | 892 | |||
BC017306.2 | - | 6 | 864 | 5088037 | ||
BX537564.1 | - | 1 | 3280 | |||
CR749209.1 | - | 2 | 4357 |
