siRNA Details
| Gene Symbol: TLR10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001017388.2 | NP_001017388.1 | 2 | 1709 | ||
| NM_001195106.1 | NP_001182035.1 | 3 | 2026 | |||
| NM_001195107.1 | NP_001182036.1 | 3 | 1898 | |||
| NM_001195108.1 | NP_001182037.1 | 3 | 1929 | |||
| NM_030956.3 | NP_112218.2 | 4 | 2215 | |||
| XM_011513760.2 | XP_011512062.1 | 2 | 1603 | |||
| XM_011513761.2 | XP_011512063.1 | 5 | 2296 | |||
| XM_011513762.2 | XP_011512064.1 | 4 | 1976 | |||
| GenBank mRNA | AB445680.1 | - | 1 | 1577 | ||
| AF296673.1 | - | 3 | 2063 | |||
| AK292026.1 | - | 2 | 1695 | |||
| AK313468.1 | - | 2 | 1846 | |||
| AY358300.1 | - | 2 | 1687 | |||
| BC089406.1 | - | 2 | 2061 | 3066588 | ||
| BC109111.2 | - | 1 | 2184 | 40035401 | ||
| BC109112.2 | - | 2 | 1727 | 40035404 | ||
| DQ011502.1 | - | 1 | 1577 | |||
| DQ011503.1 | - | 1 | 1577 | |||
| DQ011504.1 | - | 1 | 1577 | |||
| DQ011505.1 | - | 1 | 1577 | |||
| DQ011506.1 | - | 1 | 1577 | |||
| DQ011507.1 | - | 1 | 1577 | |||
| DQ011508.1 | - | 1 | 1577 | |||
| DQ011509.1 | - | 1 | 1577 | |||
| DQ011510.1 | - | 1 | 1577 | |||
| DQ011511.1 | - | 1 | 1577 | |||
| DQ011512.1 | - | 1 | 1577 | |||
| DQ011513.1 | - | 1 | 1577 | |||
| DQ011514.1 | - | 1 | 1577 | |||
| DQ011515.1 | - | 1 | 1577 | |||
| DQ011516.1 | - | 1 | 1577 | |||

TLR10
Genomic Map