siRNA Details
| Gene Symbol: SLCO5A1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001146008.1 | NP_001139480.1 | 1 | 817 | ||
| NM_001146009.1 | NP_001139481.1 | 1 | 596 | |||
| NM_030958.2 | NP_112220.2 | 2 | 807 | |||
| XM_005251313.2 | XP_005251370.1 | 2 | 807 | |||
| XM_017013883.1 | XP_016869372.1 | 2 | 807 | |||
| XM_017013884.1 | XP_016869373.1 | 2 | 807 | |||
| XM_017013886.1 | XP_016869375.1 | 2 | 807 | |||
| GenBank mRNA | AF205075.1 | - | 2 | 716 | ||
| AK022760.1 | - | 1 | 817 | |||
| AK096879.1 | - | 2 | 807 | |||
| AK299048.1 | - | 2 | 763 | |||
| AK307567.1 | - | 2 | 765 | |||
| AK308488.1 | - | 2 | 709 | |||
| BC137424.1 | - | 1 | 602 | 9021422 | ||
| BC144618.1 | - | 1 | 602 | |||

SLCO5A1
Genomic Map