siRNA Details
| Gene Symbol: OR12D3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030959.2 | NP_112221.1 | 1 | 198 | ||
| GenBank mRNA | AJ459858.1 | - | 1 | 198 | ||
| BC069082.1 | - | 1 | 194 | 7216896 | ||
| BC069154.1 | - | 1 | 194 | 7217014 | ||
| BC103892.1 | - | 1 | 198 | 40005054 | ||
| BC106898.2 | - | 1 | 991 | |||

OR12D3
Genomic Map