siRNA Details
| Gene Symbol: SPACA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030960.2 | NP_112222.1 | 2 | 345 | ||
| XM_011536160.2 | XP_011534462.1 | 2 | 299 | |||
| XM_017011335.1 | XP_016866824.1 | 2 | 295 | |||
| GenBank mRNA | AF203447.1 | - | 2 | 300 | ||
| BC029488.1 | - | 2 | 345 | 5270534 | ||

SPACA1
Genomic Map