siRNA Details
| Gene Symbol: VANGL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001172411.1 | NP_001165882.1 | 8 | 1629 | ||
| NM_001172412.1 | NP_001165883.1 | 8 | 1579 | |||
| NM_138959.2 | NP_620409.1 | 8 | 1635 | |||
| GenBank mRNA | AB057596.1 | - | 8 | 1579 | ||
| AB075805.1 | - | 7 | 1376 | |||
| AF481859.1 | - | 8 | 1591 | |||
| BC032773.1 | - | 6 | 1303 | |||
| BC065272.1 | - | 8 | 1612 | 6165098 | ||

VANGL1
Genomic Map