siRNA Details
| Gene Symbol: ZNF611 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001161499.1 | NP_001154971.1 | 6 | 2257 | ||
| NM_001161500.1 | NP_001154972.1 | 5 | 2157 | |||
| NM_001161501.1 | NP_001154973.1 | 5 | 2120 | |||
| NM_030972.3 | NP_112234.3 | 7 | 2299 | |||
| GenBank mRNA | AK021787.1 | - | 1 | 42 | ||
| AK091389.1 | - | 2 | 1837 | |||
| AK097434.1 | - | 7 | 2299 | |||
| AL834123.1 | - | 1 | 1661 | |||
| BC028590.1 | - | 9 | 2541 | |||
| BC136343.1 | - | 5 | 2205 | 9020330 | ||
| BC143996.1 | - | 4 | 2054 | |||

ZNF611
Genomic Map