siRNA Details
| Gene Symbol: PITPNM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001165966.1 | NP_001159438.1 | 17, 18 | 2531 | ||
| NM_031220.3 | NP_112497.2 | 18, 19 | 2639 | |||
| GenBank mRNA | AB209130.1 | - | 12, 13 | 5002 | ||
| AF334586.1 | - | 18, 19 | 2636 | |||
| AL389994.1 | - | 17, 18 | 2362 | |||
| BC020484.1 | - | 9, 10 | 1237 | |||
| BC128583.1 | - | 18, 19 | 2576 | 40128716 | ||
| BC128584.1 | - | 18, 19 | 2576 | 40128724 | ||

PITPNM3
Genomic Map