siRNA Details
| Gene Symbol: CDCA3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297603.1 | NP_001284532.1 | 4 | 1176 | ||
| NM_001297604.1 | NP_001284533.1 | 4 | 1176 | |||
| NM_031299.5 | NP_112589.1 | 4 | 1176 | |||
| NR_123727.1 | 4 | 1176 | ||||
| GenBank mRNA | AK092246.1 | - | 4 | 379 | ||
| AK291421.1 | - | 4 | 379 | |||
| AK312843.1 | - | 4 | 396 | |||
| BC002551.2 | - | 4 | 371 | 3138951 | ||
| BX099267.1 | - | 3 | 344 | |||
| CD106434.1 | - | 4 | 640 | |||
| DA735127.1 | - | 4 | 369 | |||

CDCA3
Genomic Map