siRNA Details
| Gene Symbol: PLVAP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031310.2 | NP_112600.1 | 1 | 130 | ||
| GenBank mRNA | AF326591.1 | - | 1 | 121 | ||
| AF348827.1 | - | 1 | 119 | |||
| AK096030.1 | - | 1 | 123 | |||
| AK313837.1 | - | 1 | 123 | |||
| AL834363.1 | - | 1 | 112 | |||
| BC050365.1 | - | 1 | 109 | 5761483 | ||
| BC056414.1 | - | 1 | 131 | 6190596 | ||
| BX380375.2 | - | 1 | 130 | |||

PLVAP
Genomic Map